missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver chromate, 99% min, Thermo Scientific Chemicals

$427.33 - $1577.82

Chemical Identifiers

CAS 7784-01-2
Molecular Formula Ag2CrO4
Molecular Weight (g/mol) 331.73
MDL Number MFCD00003402
InChI Key OJKANDGLELGDHV-UHFFFAOYSA-N
Synonym silver chromate, unii-zj5092lwu9, silver chromate vi, silberchromat, silver i chromate, 2ag.cro4, chromic acid h2cro4 , disilver 1+ salt, chromic acid h2cro4 , silver 1+ salt 1:2, chromic acid, disilver i salt, disilver dioxido dioxo chromium
PubChem CID 62666
IUPAC Name disilver;dioxido(dioxo)chromium
SMILES [O-][Cr](=O)(=O)[O-].[Ag+].[Ag+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4010714
View Documents
Thermo Scientific Chemicals
04010714
25 g
Each for $427.33
 
AA4010722
View Documents
Thermo Scientific Chemicals
04010722
100 g
Each for $1,577.82
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7784-01-2
331.73
OJKANDGLELGDHV-UHFFFAOYSA-N
62666
[O-][Cr](=O)(=O)[O-].[Ag+].[Ag+]
Ag2CrO4
MFCD00003402
silver chromate, unii-zj5092lwu9, silver chromate vi, silberchromat, silver i chromate, 2ag.cro4, chromic acid h2cro4 , disilver 1+ salt, chromic acid h2cro4 , silver 1+ salt 1:2, chromic acid, disilver i salt, disilver dioxido dioxo chromium
disilver;dioxido(dioxo)chromium

Specifications

7784-01-2
25 g
MFCD00003402
14,8510
Slightly soluble in water; Soluble in HNO3 and ammonia
[O-][Cr](=O)(=O)[O-].[Ag+].[Ag+]
331.73
331.73
Odorless
5.63 g/mL
Powder
Ag2CrO4
UN1479
silver chromate, unii-zj5092lwu9, silver chromate vi, silberchromat, silver i chromate, 2ag.cro4, chromic acid h2cro4 , disilver 1+ salt, chromic acid h2cro4 , silver 1+ salt 1:2, chromic acid, disilver i salt, disilver dioxido dioxo chromium
OJKANDGLELGDHV-UHFFFAOYSA-N
disilver;dioxido(dioxo)chromium
62666
≤99%
Light sensitive
Silver chromate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H272-H317
May cause cancer.
May intensify fire
oxidizer.
May cause an allergic skin reaction.

P201-P202-P210-P220-P221-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c

H272-H317-H350i

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

EINECSNumber : 232-043-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only