missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silver Nitrate, 0.0100 N (N/100), Ricca Chemical
ASTM for Cyanides Testing
Supplier: Ricca Chemical Company 685025
Specifications
Silver Nitrate | |
MFCD00003414 | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
ASTM for Cyanides |
AgNO3 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
Cubitainer | |
10 L |
Chemical Identifiers
AgNO3 | |
MFCD00003414 | |
silver(1+) nitrate |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
[Ag+].[O-][N+]([O-])=O |