missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silver Nitrate, 0.0100 N (N/100), Ricca Chemical
ASTM for Cyanides Testing
$84.03 - $584.69
Chemical Identifiers
Molecular Formula | AgNO3 |
---|---|
Molecular Weight (g/mol) | 169.87 |
MDL Number | MFCD00003414 |
InChI Key | SQGYOTSLMSWVJD-UHFFFAOYSA-N |
IUPAC Name | silver(1+) nitrate |
SMILES | [Ag+].[O-][N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6850-16
|
Ricca Chemical Company
685016 |
500 mL | Amber Poly Bottle |
Each for $84.03
|
|
||||
6850-32
|
Ricca Chemical Company
685032 |
1 L | Amber Poly Bottle |
Each for $110.36
|
|
||||
6850-1
|
Ricca Chemical Company
68501 |
4 L | Black Poly Bottle |
Each for $389.41
|
|
||||
6850-25
|
Ricca Chemical Company
685025 |
10 L | Cubitainer |
Each for $584.69
|
|
||||
Chemical Identifiers
AgNO3 | |
MFCD00003414 | |
silver(1+) nitrate |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
[Ag+].[O-][N+]([O-])=O |
Specifications
AgNO3 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
Amber Poly Bottle | |
500 mL |
MFCD00003414 | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
ASTM for Cyanides | |
Silver Nitrate |