missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Nitrate, 0.0100 N (N/100), Ricca Chemical

ASTM for Cyanides Testing

Supplier:  Ricca Chemical Company 685032

 View more versions of this product

Catalog No. 6850-32


Explore available promotions

Description

Description

Specifications

Specifications

SILVER NITRATE
AgNO3
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
1 L
7761-88-8
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
Amber Poly Bottle

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O
Videos
SDS
Documents

Documents

Promotions

Promotions