missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver Nitrate Solution (0.1N (N/10)/Certified), Fisher Chemical™

Standardized at 25°C against NIST™ potassium chloride

$442.67 - $7433.92

Chemical Identifiers

CAS 7761-88-8
Molecular Formula AgNO3
Molecular Weight (g/mol) 169.87
MDL Number MFCD00003414
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
Synonym silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
PubChem CID 24470
ChEBI CHEBI:32130
IUPAC Name silver(1+) nitrate
SMILES [Ag+].[O-][N+]([O-])=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
SS72-500
SDS View Documents Promotion Details
Fisher Chemical
FLSS72500
500 mL Glass Bottle
Each for $442.67
Case of 6 Each for $2,656.02
 
SS72-1 Edge
SDS View Documents Promotion Details
Fisher Chemical
FLSS721
1 L Glass Bottle
Each for $633.42
Case of 6 Each for $3,800.52
Edge
 
SS72-4 Edge
SDS Accessories View Documents
Fisher Chemical
SS724
4 L Glass Bottle
Each for $1,907.33
Case of 4 Each for $7,629.32
Edge
 
SS72-20
SDS View Documents
Fisher Chemical
FLSS7220
20 L PolyPac™
Each for $7,433.92
 
Description

Description

Specifications

Chemical Identifiers

7761-88-8
169.87
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
silver(1+) nitrate
AgNO3
MFCD00003414
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
CHEBI:32130
[Ag+].[O-][N+]([O-])=O

Specifications

Colorless
AgNO3
3082
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
[Ag+].[O-][N+]([O-])=O
169.87
CHEBI:32130
Pass Test
999b POTASSIUM CHLORIDE
500 mL
Silver Nitrate Solution, 0.1N (N/10)
7761-88-8,7732-18-5
MFCD00003414
0.0995 to 0.1005N
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver(1+) nitrate
24470
Certified
Glass Bottle
14mmHg at 20°C
Liquid
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
NFPA
Health:1 Flammability:0 Instability:0

Signal Word: CAUTION!

DOTInformation : DOT Class 5.1, : Oxidizer

SDS
missing translation for 'documents'

missing translation for 'documents'