missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silver Nitrate Solution (0.1N (N/10)/Certified), Fisher Chemical™
Standardized at 25°C against NIST™ potassium chloride
Supplier: Fisher Chemical FLSS72500
Specifications
Silver Nitrate Solution | |
0.1N (N/10) | |
AgNO3 | |
3082 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
[Ag+].[O-][N+]([O-])=O | |
169.87 | |
CHEBI:32130 | |
Pass Test | |
999b POTASSIUM CHLORIDE | |
500 mL |
Colorless | |
7761-88-8,7732-18-5 | |
MFCD00003414 | |
0.0995 to 0.1005N | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
silver(1+) nitrate | |
24470 | |
Certified | |
Glass Bottle | |
14mmHg at 20°C | |
Liquid |
Chemical Identifiers
7761-88-8 | |
169.87 | |
SQGYOTSLMSWVJD-UHFFFAOYSA-N | |
24470 | |
silver(1+) nitrate |
AgNO3 | |
MFCD00003414 | |
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol | |
CHEBI:32130 | |
[Ag+].[O-][N+]([O-])=O |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
NFPA
Health:1 Flammability:0 Instability:0
Signal Word: CAUTION!
DOTInformation : DOT Class 5.1, : Oxidizer