Simport™ Scientific SLIMSETTE™ M509 Tissue Processing/Embedding Cassettes
SLIMSETTE™ M509 Tissue Processing/Embedding Cassettes are a new generation of cassettes that are more compact and easier to use.
Supplier:  Simport™ Scientific M5093
 View more versions of this product
Catalog No. 23-045-151
Explore available promotions
Description
Description
  • Made of acetal polymer
  • Design has a unique recessed cover
  • Cassettes are thinner, allowing more to be stacked in automatic labeling machines
  • One-piece integral lid
  • Large labeling area for easy identification
  • Anterior writing area at a 45° angle
Specifications
Specifications
3 x 500/Cs.
Integral Lid
Acetal Polymer
45°
Pink
Tissue Cassette
Product Suggestions
Product Suggestions