Simport™ Scientific SLIMSETTE™ M509 Tissue Processing/Embedding Cassettes
SLIMSETTE™ M509 Tissue Processing/Embedding Cassettes are a new generation of cassettes that are more compact and easier to use.
Supplier: Simport™ Scientific M5098
Description
- Made of acetal polymer
- Design has a unique recessed cover
- Cassettes are thinner, allowing more to be stacked in automatic labeling machines
- One-piece integral lid
- Large labeling area for easy identification
- Anterior writing area at a 45° angle
Specifications
3 x 500/Cs. | |
Integral Lid | |
Acetal Polymer |
45° | |
Tan | |
Tissue Cassette |