Micro Essential Lab Single-Roll Hydrion™ pH Test Paper, With Chart
For measuring pH solutions.
Supplier:  Micro Essential Lab MES345
 View more versions of this product
Catalog No. MES345
Explore available promotions
Description
Description
  • Used in laboratories, hospitals and universities worldwide
  • Comes with a 7/32 in. x 15 ft. roll of test paper and matching color chart
  • Provides for approximately 100 tests
Specifications
Specifications
6 to 8
Colorimetric chart
pH Test Paper
For classroom demonstrations, hospital laboratories, general laboratories, industrial testing, food service, safety testing, emergency testing, agricultural testing
100 Tests