Micro Essential Lab Single-Roll Hydrion™ pH Test Paper, With Chart
For measuring pH solutions.
$18.13 - $187.33
Catalog Number | Mfr. No. | pH Range | For Use With (Application) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | pH Range | For Use With (Application) | Price | Quantity | ||||
MES345
|
Micro Essential Lab
MES345 |
6 to 8 | For classroom demonstrations, hospital laboratories, general laboratories, industrial testing, food service, safety testing, emergency testing, agricultural testing |
N/A
|
|
||||
14-853-150P
|
Micro Essential Lab
52 |
1 to 6 | For classroom demonstrations, hospital laboratories, general laboratories, industrial testing, food service, safety testing, emergency testing, agricultural testing |
N/A
|
|
||||
14-853-150C
|
Micro Essential Lab
#330 |
3.4 to 4.8 | For laboratories, hospitals and universities worldwide |
N/A
|
|
||||
14-853-150R
|
Micro Essential Lab
55 |
4 to 9 | For classroom demonstrations, hospital laboratories, general laboratories, industrial testing, food service, safety testing, emergency testing, agricultural testing |
N/A
|
|
||||
14-853-150L
|
Micro Essential Lab
#375 |
10 to 12 | For laboratories, hospitals and universities worldwide |
N/A
|
|
||||
14-853-150H
|
Micro Essential Lab
MES355 |
7.2 to 8.8 | For laboratories, hospitals and universities worldwide |
N/A
|
|
||||
14-853-150A
|
Micro Essential Lab
#305 |
0 to 3 | For classroom demonstrations, hospital laboratories, general laboratories, industrial testing, food service, safety testing, emergency testing, agricultural testing |
N/A
|
|
||||
MES-51
|
Micro Essential Lab
51 |
1 to 11 | For classroom demonstrations, hospital laboratories, general laboratories, industrial testing, food service, safety testing, emergency testing, agricultural testing |
N/A
|
|
||||
14-853-150Q
|
Micro Essential Lab
53 |
6 to 11 | For classroom demonstrations, hospital laboratories, general laboratories, industrial testing, food service, safety testing, emergency testing, agricultural testing |
N/A
|
|
||||
MES-325
|
Micro Essential Lab
MES325 |
3 to 5.5 | For classroom demonstrations, hospital laboratories, general laboratories, industrial testing, food service, safety testing, emergency testing, agricultural testing |
N/A
|
|
||||
14-853-150N
|
Micro Essential Lab
50 |
1 to 12 | For classroom demonstrations, hospital laboratories, general laboratories, industrial testing, food service, safety testing, emergency testing, agricultural testing |
N/A
|
|
||||
14-853-150G
|
Micro Essential Lab
MES350 |
6.8 to 8.4 | For laboratories, hospitals and universities worldwide |
N/A
|
|
||||
Description
- Used in laboratories, hospitals and universities worldwide
- Comes with a 7/32 in. x 15 ft. roll of test paper and matching color chart
- Provides for approximately 100 tests
Specifications
Colorimetric chart | |
100 Tests | |
pH Test Paper |