missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kord™ Slippable Petri Dishes
For standard microbiology and general laboratory use.
$294.72 - $294.72
Specifications
Disposable | Single-use |
---|---|
Material | Polystyrene |
Sterility | Sterile |
For Use With (Application) | Standard microbiology and general laboratory use |
Catalog Number | Mfr. No. | Description | Diameter (Metric) | Type | Height (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Diameter (Metric) | Type | Height (Metric) | Price | Quantity | ||
S43573
|
Kord™
2910EMD |
Featuring The Industry Standard | 100 mm | Petri Dish with Slippable Rim | 15 mm |
N/A
|
|
||
Description
- Slippable rimmed lids provide semi-stacking feature
- Deep dish design in mono style
- Polystyrene construction
- With ISO mark target
Specifications
Single-use | |
Polystyrene | |
Sterile | |
Standard microbiology and general laboratory use |