Kord™ Slippable Petri Dishes
For standard microbiology and general laboratory use.
Supplier:  Kord™ 2910EMD
 View more versions of this product
Catalog No. S43573
Explore available promotions
Description
Description
  • Slippable rimmed lids provide semi-stacking feature
  • Deep dish design in mono style
  • Polystyrene construction
  • With ISO mark target
Specifications
Specifications
Slippable
Single-use
Round
Petri Dish with Slippable Rim
Standard microbiology and general laboratory use
15 mm
Featuring The Industry Standard
Polystyrene
Sterile
1
25/Sleeve
100 mm