missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Sodium Acetate Anhydrous (White Crystals or Granular Powder), Fisher BioReagents
$91.18 - $173.90
Chemical Identifiers
CAS | 127-09-3 |
---|---|
Molecular Formula | C2H3NaO2 |
Molecular Weight (g/mol) | 82.03 |
MDL Number | MFCD00012459 |
InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
Synonym | sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech |
PubChem CID | 517045 |
ChEBI | CHEBI:32954 |
SMILES | [Na+].CC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
BP333-500
![]() |
Fisher BioReagents
BP333500 |
500 g | Poly Bottle |
Each for $91.18
Case of 6 Each for $547.08
|
![]() |
|||||
BP333-1
|
Fisher BioReagents
FLBP3331 |
1 kg | Glass Bottle |
Each for $173.90
Case of 4 Each for $695.60
|
|
|||||
Description
Sodium Acetate is added to DNA reactions to facilitate ethanol precipitation of the DNA.Chemical Identifiers
127-09-3 | |
82.03 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
517045 | |
[Na+].CC([O-])=O |
C2H3NaO2 | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
CHEBI:32954 |
Specifications
324°C | |
White | |
Powder/Solid | |
≥99.0 % | |
CH3COONa | |
15, 8709 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
sodium acetate | |
517045 | |
82.03 | |
1% max. (at 105°C) | |
Sodium Acetate Anhydrous |
127-09-3 | |
7.5 to 9.2 | |
500 g | |
C2H3NaO2 | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
[Na+].CC([O-])=O | |
82.03 | |
CHEBI:32954 | |
≥99.0% | |
Poly Bottle |
Safety and Handling

CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:1
Instability:1
Recommended Storage : RT