missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium bicarbonate, 99.5%, for analysis, Thermo Scientific Chemicals
$80.68 - $610.32
Chemical Identifiers
CAS | 144-55-8 |
---|---|
Molecular Formula | CHNaO3 |
Molecular Weight (g/mol) | 84.01 |
MDL Number | MFCD00003528 |
InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
Synonym | sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut |
PubChem CID | 516892 |
ChEBI | CHEBI:32139 |
SMILES | [Na+].OC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC217125000
|
Thermo Scientific Chemicals
217125000 |
500 g | Plastic bottle |
Each for $80.68
|
|
|||||
AC217120010
|
Thermo Scientific Chemicals
217120010 |
1 kg | Plastic bottle |
Each for $116.69
|
|
|||||
AC217120025
|
Thermo Scientific Chemicals
217120025 |
2.5 kg | Plastic bottle |
Each for $200.14
|
|
|||||
AC217120100
|
Thermo Scientific Chemicals
217120100 |
10 kg | Plastic drum |
Each for $610.32
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
144-55-8 | |
84.01 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
516892 | |
[Na+].OC([O-])=O |
CHNaO3 | |
MFCD00003528 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
CHEBI:32139 |
Specifications
270.0°C | |
White | |
500 g | |
CHNaO3 | |
MFCD00003528 | |
15, 8719 | |
Solubility in water: 9 g/100mL (20°C). Other solubilities: 16.4 g/100mL water (60°C), insoluble in alcohol | |
[Na+].OC([O-])=O | |
84.01 | |
CHEBI:32139 | |
99.5% | |
Plastic bottle |
144-55-8 | |
Fine Crystalline Powder | |
99.50% | |
NaHCO3 | |
03,260; 05,595; 11,476; 17,180 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
sodium;hydrogen carbonate | |
516892 | |
84.01 | |
Analytical | |
Sodium bicarbonate |
Safety and Handling
EINECSNumber : 205-633-8
RUO – Research Use Only