missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium bis(trimethylsilyl)amide, 95+%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 215961000

 View more versions of this product

Catalog No. AC215961000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium bis(trimethylsilyl)amide
171°C to 175°C
.916g/mL
95+%
C6H18NNaSi2
MFCD00009835
01,1046; 03,261; 04,442; 06,529; 12,441; 16,307
Solubility in water: reacts
C[Si](C)(C)[N-][Si](C)(C)C.[Na+]
183.38
183.38
Pure
1070-89-9
Yellow to Beige
Authentic
Glass bottle
[(CH3)3Si]2NNa
100 g
sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt
WRIKHQLVHPKCJU-UHFFFAOYSA-N
sodium;bis(trimethylsilyl)azanide
2724254
95+%
Crystalline Powder

Chemical Identifiers

1070-89-9
183.38
WRIKHQLVHPKCJU-UHFFFAOYSA-N
2724254
C[Si](C)(C)[N-][Si](C)(C)C.[Na+]
C6H18NNaSi2
MFCD00009835
sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt
sodium;bis(trimethylsilyl)azanide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Flammable solid.
Reacts violently with water.

GHS P Statement
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiou

GHS Signal Word: Danger

EINECSNumber : 213-983-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only