Learn More
Sodium bis(trimethylsilyl)amide, 95+%, pure, Thermo Scientific Chemicals
$234.20 - $566.53
Chemical Identifiers
CAS | 1070-89-9 |
---|---|
Molecular Formula | C6H18NNaSi2 |
Molecular Weight (g/mol) | 183.38 |
MDL Number | MFCD00009835 |
InChI Key | WRIKHQLVHPKCJU-UHFFFAOYSA-N |
Synonym | sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt |
PubChem CID | 2724254 |
IUPAC Name | sodium;bis(trimethylsilyl)azanide |
SMILES | C[Si](C)(C)[N-][Si](C)(C)C.[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC215960250
|
Thermo Scientific Chemicals
215960250 |
25 g | Glass bottle |
Each for $234.20
|
|
||||
AC215961000
|
Thermo Scientific Chemicals
215961000 |
100 g | Glass bottle |
Each for $566.53
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1070-89-9 | |
183.38 | |
WRIKHQLVHPKCJU-UHFFFAOYSA-N | |
2724254 | |
C[Si](C)(C)[N-][Si](C)(C)C.[Na+] |
C6H18NNaSi2 | |
MFCD00009835 | |
sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt | |
sodium;bis(trimethylsilyl)azanide |
Specifications
1070-89-9 | |
Yellow to Beige | |
Authentic | |
Glass bottle | |
[(CH3)3Si]2NNa | |
25 g | |
sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt | |
WRIKHQLVHPKCJU-UHFFFAOYSA-N | |
sodium;bis(trimethylsilyl)azanide | |
2724254 | |
95+% | |
Crystalline Powder |
171°C to 175°C | |
.916g/mL | |
95+% | |
C6H18NNaSi2 | |
MFCD00009835 | |
01,1046; 03,261; 04,442; 06,529; 12,441; 16,307 | |
Solubility in water: reacts | |
C[Si](C)(C)[N-][Si](C)(C)C.[Na+] | |
183.38 | |
183.38 | |
Pure | |
Sodium bis(trimethylsilyl)amide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Flammable solid.
Reacts violently with water.
GHS P Statement
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiou
GHS Signal Word: Danger
EINECSNumber : 213-983-8
RUO – Research Use Only