missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 277851000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium bis(trimethylsilyl)amide | |
2M Solution in THF | |
C6H18NNaSi2 | |
MFCD00009835 | |
0.9160g/mL | |
WRIKHQLVHPKCJU-UHFFFAOYSA-N | |
sodium;bis(trimethylsilyl)azanide | |
2724254 | |
38 to 42% | |
AcroSeal™ Glass bottle | |
100 mL | |
Liquid |
Orange to Brown | |
1070-89-9,109-99-9 | |
[(CH3)3Si]2NNa | |
01,1046; 03,261; 04,442; 06,529; 12,441; 16,307 | |
sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt | |
C[Si](C)(C)[N-][Si](C)(C)C.[Na+] | |
183.377 | |
183.38 | |
Pure | |
0.916 | |
−21°C |
RUO – Research Use Only