missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals

$90.06 - $419.15

Specifications

Chemical Name or Material Sodium bis(trimethylsilyl)amide
Color Orange to Brown
Name Note 2M Solution in THF
CAS 1070-89-9,109-99-9
Molecular Formula C6H18NNaSi2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC277851000
View Documents
Thermo Scientific Chemicals
277851000
100 mL AcroSeal™ Glass bottle
Each for $90.06
 
AC277858000
View Documents
Thermo Scientific Chemicals
277858000
800 mL AcroSeal™ Glass Bottle
Each for $419.15
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium bis(trimethylsilyl)amide
2M Solution in THF
C6H18NNaSi2
MFCD00009835
0.9160g/mL
WRIKHQLVHPKCJU-UHFFFAOYSA-N
sodium;bis(trimethylsilyl)azanide
2724254
38 to 42%
0.916
Liquid
Orange to Brown
1070-89-9,109-99-9
[(CH3)3Si]2NNa
01,1046; 03,261; 04,442; 06,529; 12,441; 16,307
sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt
C[Si](C)(C)[N-][Si](C)(C)C.[Na+]
183.377
183.38
Pure
−21°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only