missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Carbonate Anhydrous (Powder/Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLS26310

 View more versions of this product

Catalog No. S263-10


Explore available promotions

Description

Description

Specifications

Specifications

1600°C
854°C
White
Solid
≥99.5 %
CNa2O3
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
[Na+].[Na+].[O-]C([O-])=O
105.99
CHEBI:29377
≥99.5%
Pass Test
2.53g/cm³
Sodium Carbonate Anhydrous
497-19-8
11.3
10 kg
(dried basis)≥99.5%
MFCD00003494
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium carbonate
10340
105.99
Certified ACS
Poly Pail

Chemical Identifiers

497-19-8
105.99
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
disodium carbonate
CNa2O3
MFCD00003494
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
CHEBI:29377
[Na+].[Na+].[O-]C([O-])=O
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Irritating to eyes and skin. May cause central nervous system effects. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Use personal protective equipment. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:0
Instability:1

SDS Documents

Documents

Promotions

Promotions