missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Carbonate Anhydrous (Powder/Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical FLS26350
Specifications
1600°C | |
854°C | |
White | |
Solid | |
≥99.5% | |
CNa2O3 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
[Na+].[Na+].[O-]C([O-])=O | |
105.99 | |
CHEBI:29377 | |
≥99.5% | |
Pass Test | |
2.53g/cm³ |
Sodium Carbonate Anhydrous | |
497-19-8 | |
11.3 | |
50 kg | |
(dried basis)≥99.5% | |
MFCD00003494 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
disodium carbonate | |
10340 | |
105.99 | |
Certified ACS | |
Fiber Drum |
Chemical Identifiers
497-19-8 | |
105.99 | |
CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
10340 | |
disodium carbonate |
CNa2O3 | |
MFCD00003494 | |
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
CHEBI:29377 | |
[Na+].[Na+].[O-]C([O-])=O |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes and skin. May cause central nervous system effects. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Do not get in eyes, on skin, or on clothing. Use personal protective equipment. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.
NFPA
Health:2
Flammability:0
Instability:1