missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium carbonate decahydrate, 99+%, extra pure, Thermo Scientific Chemicals

$30.86 - $384.59

Chemical Identifiers

CAS 2-1-6132
Molecular Formula CNa2O3·10H2O
Molecular Weight (g/mol) 286.14
InChI Key XYQRXRFVKUPBQN-UHFFFAOYSA-L
Synonym sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
PubChem CID 151402
IUPAC Name disodium;carbonate;decahydrate
SMILES C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC271071000
SDS View Documents
Thermo Scientific Chemicals
271071000
100 g Plastic Bottle
Each for $30.86
 
AC271070100
View Documents
Thermo Scientific Chemicals
271070100
10 kg Plastic Drum
Each for $384.59
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2-1-6132
286.14
sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
disodium;carbonate;decahydrate
CNa2O3·10H2O
XYQRXRFVKUPBQN-UHFFFAOYSA-L
151402
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]

Specifications

34.0°C
White
100 g
CNa2O3·10H2O
7322
sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t
XYQRXRFVKUPBQN-UHFFFAOYSA-L
disodium;carbonate;decahydrate
151402
≥99%
Plastic Bottle
1.46 g/mL
2-1-6132
Crystals
99+%
Na2CO3·10H2O
158731
Solubility in water: 210g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]
286.14
286.14
Extra Pure
1.46
Sodium carbonate decahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only