missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium cyanoborohydride, 95%, Thermo Scientific Chemicals

Selective Reducing Agent For Aldehydes, Ketones, Oximes

Supplier:  Thermo Scientific Chemicals 08783922

 View more versions of this product

Catalog No. AA8783922


Explore available promotions

Description

Description

Selective reducing agent for aldehydes, ketones, oximes, enamines. Reductive animation of ketones, aldehydes. Reductive alkylation of amines, hydrazines Sodium cyanoborohydride is used as a reagent involved in the reductive amination of aldehydes, ketones, oximes and enamines. It is also used in the reductive alkylation of amines and hydrazines. It acts as a catalyst in the preparation of phenolate-bridged dilanthanum(III) complex. In addition, it is used in the conversion of imines to amines. It plays an important role in the reductive deoxygenation of ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Moisture Sensitive; Highly flammable
Specifications

Specifications

Sodium cyanoborohydride
25895-60-7
Powder
95%
MFCD00003516
Soluble in water,THF,methanolSoluble in water,diglyme,tetrahydrofuran and methanol. Slightly soluble in dimethyl sulfoxide. Insoluble in diethyl ether.
[Na+].[BH3-]C#N
62.84
62.84
1.2
>300°C (decomposition)
White
100 g
CH3BNNa
sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
CVDUGUOQTVTBJH-UHFFFAOYSA-N
sodium cyanoboranuide
20587905
Moisture sensitive

Chemical Identifiers

25895-60-7
62.84
CVDUGUOQTVTBJH-UHFFFAOYSA-N
20587905
CH3BNNa
MFCD00003516
sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
[Na+].[BH3-]C#N
Videos
Safety and Handling

Safety and Handling

P210-P223-P231+P232-P240-P241-P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c

H228-H261-H300+H310+H330-H314

DOTInformation : DOT Class: 4.3, Packaging Group: II

EINECSNumber : 247-317-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only