missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium dihydrogen citrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1255736

 View more versions of this product

Catalog No. AAL1255736


Explore available promotions

Description

Description

Sodium dihydrogen citrate acts as a buffering agent and sequestrant. It is also employed as a reductant and stabilizer in the preparation of b>Fe3O4 platinum core/shell particles. Further, it is used in electrochemical reduction of proton at carbon fiber electrode in the presence and absence of platinum nanoparticles. In addition to this, it plays a useful role in characterization of carbon-based electrode materials as cathode in half-cell configuration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dihydrogen citrate acts as a buffering agent and sequestrant. It is also employed as a reductant and stabilizer in the preparation of b>Fe3O4 platinum core/shell particles. Further, it is used in electrochemical reduction of proton at carbon fiber electrode in the presence and absence of platinum nanoparticles. In addition to this, it plays a useful role in characterization of carbon-based electrode materials as cathode in half-cell configuration.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Hygroscopic. Incompatible with bases, reducing agents and oxidizing agents.
Specifications

Specifications

Sodium dihydrogen citrate
211°C to 212°C
MFCD00013067
3920246
Soluble in water. Insoluble in ethanol.
[HH].C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[Na]
217.129
214.11
18996-35-5
C6H10NaO7
500 g
bicitra, pneucid, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt, sodium dihydrogen citrate anhydrous, citric acid, sodium salt, citric acid sodium, sodium 2-hydroxy-1,2,3-propanetricarboxylate, 2-hydroxypropane-1,2,3-tricarboxylic acid; sodium, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt 1:?
SLWOXBQHKZPUNY-UHFFFAOYSA-N
2-hydroxypropane-1,2,3-tricarboxylic acid;molecular hydrogen;sodium
131675399
99%

Chemical Identifiers

18996-35-5
217.129
SLWOXBQHKZPUNY-UHFFFAOYSA-N
131675399
[HH].C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[Na]
C6H10NaO7
MFCD00013067
bicitra, pneucid, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt, sodium dihydrogen citrate anhydrous, citric acid, sodium salt, citric acid sodium, sodium 2-hydroxy-1,2,3-propanetricarboxylate, 2-hydroxypropane-1,2,3-tricarboxylic acid; sodium, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt 1:?
2-hydroxypropane-1,2,3-tricarboxylic acid;molecular hydrogen;sodium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P264b-P280i-P305+P351+P338

H319-H500

EINECSNumber : 242-734-6

RTECSNumber : GE7950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only