Learn More
Sodium dihydrogen citrate, 99%, Thermo Scientific Chemicals
$63.74 - $290.36
Chemical Identifiers
CAS | 18996-35-5 |
---|---|
Molecular Formula | C6H10NaO7 |
Molecular Weight (g/mol) | 217.129 |
MDL Number | MFCD00013067 |
InChI Key | SLWOXBQHKZPUNY-UHFFFAOYSA-N |
Synonym | bicitra, pneucid, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt, sodium dihydrogen citrate anhydrous, citric acid, sodium salt, citric acid sodium, sodium 2-hydroxy-1,2,3-propanetricarboxylate, 2-hydroxypropane-1,2,3-tricarboxylic acid; sodium, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt 1:? |
PubChem CID | 131675399 |
IUPAC Name | 2-hydroxypropane-1,2,3-tricarboxylic acid;molecular hydrogen;sodium |
SMILES | [HH].C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[Na] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1255722
|
Thermo Scientific Chemicals
L1255722 |
100 g |
Each for $63.74
|
|
|||||
AAL1255736
|
Thermo Scientific Chemicals
L1255736 |
500 g |
Each for $115.22
|
|
|||||
AAL125570P
|
Thermo Scientific Chemicals
L125570P |
2000 g |
Each for $290.36
|
|
|||||
Description
Sodium dihydrogen citrate acts as a buffering agent and sequestrant. It is also employed as a reductant and stabilizer in the preparation of b>Fe3O4 platinum core/shell particles. Further, it is used in electrochemical reduction of proton at carbon fiber electrode in the presence and absence of platinum nanoparticles. In addition to this, it plays a useful role in characterization of carbon-based electrode materials as cathode in half-cell configuration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium dihydrogen citrate acts as a buffering agent and sequestrant. It is also employed as a reductant and stabilizer in the preparation of Fe3O4 platinum core/shell particles. Further, it is used in electrochemical reduction of proton at carbon fiber electrode in the presence and absence of platinum nanoparticles. In addition to this, it plays a useful role in characterization of carbon-based electrode materials as cathode in half-cell configuration.
Solubility
Soluble in water. Insoluble in ethanol.
Notes
Hygroscopic. Incompatible with bases, reducing agents and oxidizing agents.
Chemical Identifiers
18996-35-5 | |
217.129 | |
SLWOXBQHKZPUNY-UHFFFAOYSA-N | |
131675399 | |
[HH].C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[Na] |
C6H10NaO7 | |
MFCD00013067 | |
bicitra, pneucid, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt, sodium dihydrogen citrate anhydrous, citric acid, sodium salt, citric acid sodium, sodium 2-hydroxy-1,2,3-propanetricarboxylate, 2-hydroxypropane-1,2,3-tricarboxylic acid; sodium, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt 1:? | |
2-hydroxypropane-1,2,3-tricarboxylic acid;molecular hydrogen;sodium |
Specifications
18996-35-5 | |
C6H10NaO7 | |
100 g | |
bicitra, pneucid, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt, sodium dihydrogen citrate anhydrous, citric acid, sodium salt, citric acid sodium, sodium 2-hydroxy-1,2,3-propanetricarboxylate, 2-hydroxypropane-1,2,3-tricarboxylic acid; sodium, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt 1:? | |
SLWOXBQHKZPUNY-UHFFFAOYSA-N | |
2-hydroxypropane-1,2,3-tricarboxylic acid;molecular hydrogen;sodium | |
131675399 | |
99% |
211°C to 212°C | |
MFCD00013067 | |
3920246 | |
Soluble in water. Insoluble in ethanol. | |
[HH].C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[Na] | |
217.129 | |
214.11 | |
Sodium dihydrogen citrate |
Safety and Handling
P264b-P280i-P305+P351+P338
H319-H500
EINECSNumber : 242-734-6
RTECSNumber : GE7950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only