missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium dodecyl sulfate, ≥99.5% alkyl sulfate, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219895780

 View more versions of this product

Catalog No. ICN19895780


Explore available promotions

Description

Description

Specifications

Specifications

Sodium dodecyl sulfate, alkyl sulfate
151-21-3
100 mL
C12H25NaO4S
Sodium lauryl sulfate, Sulfuric acid
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
CHEBI:8984
≥99.5% (alkyl sulfate)
Colorless
Liquid
>99.5%
MFCD00036175
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium;dodecyl sulfate
3423265
288.38

Chemical Identifiers

151-21-3
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
3423265
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
C12H25NaO4S
MFCD00036175
Sodium lauryl sulfate, Sulfuric acid
CHEBI:8984
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-791-2

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Promotions

Promotions