missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium dodecyl sulfate, ≥99.5% alkyl sulfate, MP Biomedicals™
$119.93 - $448.49
Chemical Identifiers
CAS | 151-21-3 |
---|---|
Molecular Formula | C12H25NaO4S |
Molecular Weight (g/mol) | 288.38 |
MDL Number | MFCD00036175 |
InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
Synonym | Sodium lauryl sulfate, Sulfuric acid |
PubChem CID | 3423265 |
ChEBI | CHEBI:8984 |
SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN19895780
|
MP Biomedicals Inc
0219895780 |
100 mL |
Each for $119.93
|
|
|||||
ICN19895790
|
MP Biomedicals Inc
0219895790 |
500 mL |
Each for $448.49
|
|
|||||
Chemical Identifiers
151-21-3 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
3423265 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
C12H25NaO4S | |
MFCD00036175 | |
Sodium lauryl sulfate, Sulfuric acid | |
CHEBI:8984 |
Specifications
Colorless | |
Liquid | |
>99.5% | |
MFCD00036175 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium;dodecyl sulfate | |
3423265 | |
288.38 | |
Sodium dodecyl sulfate, alkyl sulfate |
151-21-3 | |
100 mL | |
C12H25NaO4S | |
Sodium lauryl sulfate, Sulfuric acid | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
CHEBI:8984 | |
≥99.5% (alkyl sulfate) |
Safety and Handling
EINECSNumber : 231-791-2
Recommended Storage : Store at room temperature.