missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Dodecyl Sulfate (SDS), Micropellets, Fisher BioReagents
Sodium Dodecyl Sulfate is the most commonly used detergent in protein purification and electrophoresis
Supplier: Thermo Fisher Scientific BP8200100
Specifications
≤0.10 (0.1% Solution) at 260nm | |
White | |
Solid | |
>99% | |
MFCD00036175 | |
100 g | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
3423265 | |
≥98.0% (C12 alcohol) | |
Poly Bottle |
Sodium Dodecyl Sulfate (SDS), Micropellets | |
151-21-3 | |
204°C | |
C12H25NaO4S | |
8.5 to 10 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium;dodecyl sulfate | |
Pass test | |
CHEBI:8984 | |
Molecular Biology |
Chemical Identifiers
151-21-3 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
CHEBI:8984 |
C12H25NaO4S | |
MFCD00036175 | |
3423265 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Safety and Handling
Recommended Storage : Store at room temperature