missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Dodecyl Sulfate (SDS), Micropellets, Fisher BioReagents
Sodium Dodecyl Sulfate is the most commonly used detergent in protein purification and electrophoresis
$380.21 - $5106.00
Chemical Identifiers
CAS | 151-21-3 |
---|---|
Molecular Formula | C12H25NaO4S |
Molecular Weight (g/mol) | 288.38 |
MDL Number | MFCD00036175 |
InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
PubChem CID | 3423265 |
ChEBI | CHEBI:8984 |
SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP8200100
|
Thermo Fisher Scientific
BP8200100 |
100 g | Poly Bottle |
Each for $380.21
|
|
||||
BP8200500
|
Thermo Fisher Scientific
BP8200500 |
500 g | Poly Bottle |
Each for $982.26
|
|
||||
BP82005
|
Thermo Fisher Scientific
BP82005 |
5 kg | Poly Pail |
Each for $5,106.00
|
|
||||
Chemical Identifiers
151-21-3 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
CHEBI:8984 |
C12H25NaO4S | |
MFCD00036175 | |
3423265 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Specifications
≤0.10 (0.1% Solution) at 260nm | |
151-21-3 | |
204°C | |
C12H25NaO4S | |
8.5 to 10 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium;dodecyl sulfate | |
Pass test | |
CHEBI:8984 | |
Molecular Biology | |
Sodium Dodecyl Sulfate (SDS), Micropellets |
White | |
Solid | |
>99% | |
MFCD00036175 | |
100 g | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
3423265 | |
≥98.0% (C12 alcohol) | |
Poly Bottle |
Safety and Handling
Recommended Storage : Store at room temperature