missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Ferrocyanide Decahydrate, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1274500GM

 View more versions of this product

Catalog No. 18613481


Explore available promotions

Description

Description

Specifications

Specifications

14434-22-1
500 g
Reagent
1
99%
Amber Glass Bottle

Chemical Identifiers

14434-22-1
Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.