missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Ferrocyanide Decahydrate, 99%, Spectrum™ Chemical

$757.12 - $24293.76

Chemical Identifiers

CAS 14434-22-1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613481
View Documents
Spectrum Chemical Mfg Cor
S1274500GM
500 g Amber Glass Bottle
Each for $757.12
 
18613482
View Documents
Spectrum Chemical Mfg Cor
S127425KG
2.5 kg Amber Glass Bottle
Each for $2,044.41
 
18613483
View Documents
Spectrum Chemical Mfg Cor
S127412KG
12 kg Poly Pail
Each for $6,895.29
 
18613484
View Documents
Spectrum Chemical Mfg Cor
S127450KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

14434-22-1

Specifications

14434-22-1
500 g
Reagent
1
99%
Amber Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.