missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hexafluorophosphate, 98.5+%, pure, Thermo Scientific Chemicals

Sodium hexafluorophosphate, 98.5+%, pure, Acros Organics, Quantity: 50g, Packaging: Plastic bottle, CAS: 21324-39-0, White to Gray, Grade: Pure, Melting Point: 200.0 deg.C, Molecular Formula: F6NaP, Molecular Weight: 167.95, Percent Purity: 98.5+%

Supplier:  Thermo Scientific Chemicals 196820100

 View more versions of this product

Catalog No. AC196820100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Specifications

Sodium hexafluorophosphate
21324-39-0
Crystalline Powder
98.5% min
NaPF6
sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1
KMADQUOCJBLXRP-UHFFFAOYSA-N
sodium hexafluoro-λ⁵-phosphanuide
5147921
≥98.5%
Plastic bottle
200.0°C
White to Gray
10 g
F6NaP
MFCD00011122
Solubility in water: soluble
[Na+].F[P-](F)(F)(F)(F)F
167.95
167.95
Pure

Chemical Identifiers

21324-39-0
167.95
KMADQUOCJBLXRP-UHFFFAOYSA-N
5147921
[Na+].F[P-](F)(F)(F)(F)F
F6NaP
MFCD00011122
sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1
sodium hexafluoro-λ⁵-phosphanuide
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

EINECSNumber : 244-333-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only