Learn More
Sodium hexafluorophosphate, 98.5+%, pure, Thermo Scientific Chemicals
Sodium hexafluorophosphate, 98.5+%, pure, Acros Organics, Quantity: 50g, Packaging: Plastic bottle, CAS: 21324-39-0, White to Gray, Grade: Pure, Melting Point: 200.0 deg.C, Molecular Formula: F6NaP, Molecular Weight: 167.95, Percent Purity: 98.5+%
$85.96 - $1133.53
Chemical Identifiers
CAS | 21324-39-0 |
---|---|
Molecular Formula | F6NaP |
Molecular Weight (g/mol) | 167.95 |
MDL Number | MFCD00011122 |
InChI Key | KMADQUOCJBLXRP-UHFFFAOYSA-N |
Synonym | sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1 |
PubChem CID | 5147921 |
IUPAC Name | sodium hexafluoro-λâµ-phosphanuide |
SMILES | [Na+].F[P-](F)(F)(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC196820100
|
Thermo Scientific Chemicals
196820100 |
10 g | Plastic bottle |
Each for $85.96
|
|
||||
AC196820500
|
Thermo Scientific Chemicals
196820500 |
50 g | Plastic bottle |
Each for $329.55
|
|
||||
AC196822500
|
Thermo Scientific Chemicals
196822500 |
250 g | Plastic bottle |
Each for $1,133.53
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
21324-39-0 | |
167.95 | |
KMADQUOCJBLXRP-UHFFFAOYSA-N | |
5147921 | |
[Na+].F[P-](F)(F)(F)(F)F |
F6NaP | |
MFCD00011122 | |
sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1 | |
sodium hexafluoro-λâµ-phosphanuide |
Specifications
200.0°C | |
White to Gray | |
10 g | |
F6NaP | |
MFCD00011122 | |
Solubility in water: soluble | |
[Na+].F[P-](F)(F)(F)(F)F | |
167.95 | |
167.95 | |
Pure | |
Sodium hexafluorophosphate |
21324-39-0 | |
Crystalline Powder | |
98.5% min | |
NaPF6 | |
sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1 | |
KMADQUOCJBLXRP-UHFFFAOYSA-N | |
sodium hexafluoro-λâµ-phosphanuide | |
5147921 | |
≥98.5% | |
Plastic bottle |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 244-333-1
RUO – Research Use Only