Learn More
Sodium hexafluorophosphate, 98.5+%, pure, Thermo Scientific Chemicals
Sodium hexafluorophosphate, 98.5+%, pure, Acros Organics, Quantity: 50g, Packaging: Plastic bottle, CAS: 21324-39-0, White to Gray, Grade: Pure, Melting Point: 200.0 deg.C, Molecular Formula: F6NaP, Molecular Weight: 167.95, Percent Purity: 98.5+%
Supplier: Thermo Scientific Chemicals 196820500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Sodium hexafluorophosphate | |
21324-39-0 | |
Crystalline Powder | |
98.5% min | |
NaPF6 | |
sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1 | |
KMADQUOCJBLXRP-UHFFFAOYSA-N | |
sodium hexafluoro-λâµ-phosphanuide | |
5147921 | |
≥98.5% | |
Plastic bottle |
200.0°C | |
White to Gray | |
50 g | |
F6NaP | |
MFCD00011122 | |
Solubility in water: soluble | |
[Na+].F[P-](F)(F)(F)(F)F | |
167.95 | |
167.95 | |
Pure |
Chemical Identifiers
21324-39-0 | |
167.95 | |
KMADQUOCJBLXRP-UHFFFAOYSA-N | |
5147921 | |
[Na+].F[P-](F)(F)(F)(F)F |
F6NaP | |
MFCD00011122 | |
sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1 | |
sodium hexafluoro-λâµ-phosphanuide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 244-333-1
RUO – Research Use Only