missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hexanoate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 437540250

 View more versions of this product

Catalog No. AC437540250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium hexanoate
White
Powder
99%
MFCD00059056
UDWXLZLRRVQONG-UHFFFAOYSA-M
sodium;hexanoate
4087444
99%
2% max. ()
231.2°C to 233.8°C
10051-44-2
25 g
C6H11NaO2
sodium hexanoate, sodium caproate, hexanoic acid, sodium salt, hexanoic acid sodium salt, sodium capronate, sodium n-caproate, sodium n-hexanoate, unii-e00cg11s66, caproic acid sodium salt, hexanoic acid, sodium salt 1:1
[Na+].CCCCCC([O-])=O
138.14
138.14
Authentic

Chemical Identifiers

10051-44-2
138.14
UDWXLZLRRVQONG-UHFFFAOYSA-M
4087444
[Na+].CCCCCC([O-])=O
C6H11NaO2
MFCD00059056
sodium hexanoate, sodium caproate, hexanoic acid, sodium salt, hexanoic acid sodium salt, sodium capronate, sodium n-caproate, sodium n-hexanoate, unii-e00cg11s66, caproic acid sodium salt, hexanoic acid, sodium salt 1:1
sodium;hexanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 233-179-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only