missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hexanoate, 99%, Thermo Scientific Chemicals

$170.73 - $338.79

Chemical Identifiers

CAS 10051-44-2
Molecular Formula C6H11NaO2
Molecular Weight (g/mol) 138.14
MDL Number MFCD00059056
InChI Key UDWXLZLRRVQONG-UHFFFAOYSA-M
Synonym sodium hexanoate, sodium caproate, hexanoic acid, sodium salt, hexanoic acid sodium salt, sodium capronate, sodium n-caproate, sodium n-hexanoate, unii-e00cg11s66, caproic acid sodium salt, hexanoic acid, sodium salt 1:1
PubChem CID 4087444
IUPAC Name sodium;hexanoate
SMILES [Na+].CCCCCC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC437540100
SDS View Documents
Thermo Scientific Chemicals
437540100
10 g
Each for $170.73
 
AC437540250
SDS View Documents
Thermo Scientific Chemicals
437540250
25 g
Each for $338.79
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10051-44-2
138.14
UDWXLZLRRVQONG-UHFFFAOYSA-M
4087444
[Na+].CCCCCC([O-])=O
C6H11NaO2
MFCD00059056
sodium hexanoate, sodium caproate, hexanoic acid, sodium salt, hexanoic acid sodium salt, sodium capronate, sodium n-caproate, sodium n-hexanoate, unii-e00cg11s66, caproic acid sodium salt, hexanoic acid, sodium salt 1:1
sodium;hexanoate

Specifications

231.2°C to 233.8°C
10051-44-2
10 g
C6H11NaO2
sodium hexanoate, sodium caproate, hexanoic acid, sodium salt, hexanoic acid sodium salt, sodium capronate, sodium n-caproate, sodium n-hexanoate, unii-e00cg11s66, caproic acid sodium salt, hexanoic acid, sodium salt 1:1
[Na+].CCCCCC([O-])=O
138.14
138.14
Authentic
Glass Bottle
White
Powder
99%
MFCD00059056
UDWXLZLRRVQONG-UHFFFAOYSA-M
sodium;hexanoate
4087444
99%
2% max. ()
Sodium hexanoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 233-179-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only