missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 251770010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium hyaluronate | |
White to Cream | |
10% max. (500mg, 105°C, 6 hrs) | |
94% min. (On dry substance) | |
(C14H20NO11Na)n | |
MFCD00875848 | |
15,4796 | |
Solubility in water: soluble. | |
CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-* | |
417.30 | |
Powder |
9067-32-7 | |
6.0 to 8.0 (0.5% aq. soln.) | |
Authentic | |
Glass bottle | |
(C14H20NO11Na)n | |
1 g | |
hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid | |
YWIVKILSMZOHHF-QJZPQSOGSA-N | |
Sodium hyaluronate | |
95% |
Chemical Identifiers
9067-32-7 | |
417.30 | |
YWIVKILSMZOHHF-QJZPQSOGSA-N | |
Sodium hyaluronate |
(C14H20NO11Na)n | |
MFCD00875848 | |
hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid | |
CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-* |
RUO – Research Use Only