missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
$152.51 - $489.46
Chemical Identifiers
CAS | 9067-32-7 |
---|---|
Molecular Formula | (C14H20NO11Na)n |
Molecular Weight (g/mol) | 417.30 |
MDL Number | MFCD00875848 |
InChI Key | YWIVKILSMZOHHF-QJZPQSOGSA-N |
Synonym | hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
IUPAC Name | Sodium hyaluronate |
SMILES | CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-* |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC251770010
|
Thermo Scientific Chemicals
251770010 |
1 g | Glass bottle |
Each for $152.51
|
|
||||
AC251770050
|
Thermo Scientific Chemicals
251770050 |
5 g | Glass bottle |
Each for $489.46
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9067-32-7 | |
417.30 | |
YWIVKILSMZOHHF-QJZPQSOGSA-N | |
Sodium hyaluronate |
(C14H20NO11Na)n | |
MFCD00875848 | |
hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid | |
CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-* |
Specifications
9067-32-7 | |
6.0 to 8.0 (0.5% aq. soln.) | |
Authentic | |
Glass bottle | |
(C14H20NO11Na)n | |
1 g | |
hyaluronate tetrasaccharide, d0e9sz, 6-3-acetamido-2-6-3-acetamido-2,5-dihydroxy-6-hydroxymethyl oxan-4-yl oxy-2-carboxy-4,5-dihydroxyoxan-3-yl oxy-5-hydroxy-6-hydroxymethyl oxan-4-yl oxy-3,4,5-trihydroxyoxane-2-carboxylic acid | |
YWIVKILSMZOHHF-QJZPQSOGSA-N | |
Sodium hyaluronate | |
95% | |
Sodium hyaluronate |
White to Cream | |
10% max. (500mg, 105°C, 6 hrs) | |
94% min. (On dry substance) | |
(C14H20NO11Na)n | |
MFCD00875848 | |
15,4796 | |
Solubility in water: soluble. | |
CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-* | |
417.30 | |
Powder |
RUO – Research Use Only