missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium hydrogen DL-malate, 98%, Thermo Scientific Chemicals
$98.12 - $320.38
Chemical Identifiers
CAS | 57467-17-1 |
---|---|
Molecular Formula | C4H5NaO5 |
Molecular Weight (g/mol) | 156.07 |
MDL Number | MFCD00798569 |
InChI Key | DOJOZCIMYABYPO-UHFFFAOYNA-M |
Synonym | sodium hydrogen malate, monosodium malate, sodium malate, l-malic acid, --sodium hydrogen malate, l--malic acid disodium salt, sodium 3-carboxy-3-hydroxypropanoate, 2-hydroxysuccinic acid 1-sodium salt, sodium 3,4-dihydroxy-4-oxo-butanoate, 2-hydroxybutanedioic acid 4-sodium salt |
PubChem CID | 23678821 |
IUPAC Name | sodium;3,4-dihydroxy-4-oxobutanoate |
SMILES | [Na+].OC(CC([O-])=O)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1942818
|
Thermo Scientific Chemicals
A1942818 |
50 g |
Each for $98.12
|
|
|||||
AAA1942830
|
Thermo Scientific Chemicals
A1942830 |
250 g |
Each for $320.38
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
57467-17-1 | |
156.07 | |
DOJOZCIMYABYPO-UHFFFAOYNA-M | |
23678821 | |
[Na+].OC(CC([O-])=O)C(O)=O |
C4H5NaO5 | |
MFCD00798569 | |
sodium hydrogen malate, monosodium malate, sodium malate, l-malic acid, --sodium hydrogen malate, l--malic acid disodium salt, sodium 3-carboxy-3-hydroxypropanoate, 2-hydroxysuccinic acid 1-sodium salt, sodium 3,4-dihydroxy-4-oxo-butanoate, 2-hydroxybutanedioic acid 4-sodium salt | |
sodium;3,4-dihydroxy-4-oxobutanoate |
Specifications
57467-17-1 | |
C4H5NaO5 | |
50 g | |
DOJOZCIMYABYPO-UHFFFAOYNA-M | |
sodium;3,4-dihydroxy-4-oxobutanoate | |
23678821 | |
98% |
210°C (decomposition) | |
MFCD00798569 | |
sodium hydrogen malate, monosodium malate, sodium malate, l-malic acid, --sodium hydrogen malate, l--malic acid disodium salt, sodium 3-carboxy-3-hydroxypropanoate, 2-hydroxysuccinic acid 1-sodium salt, sodium 3,4-dihydroxy-4-oxo-butanoate, 2-hydroxybutanedioic acid 4-sodium salt | |
[Na+].OC(CC([O-])=O)C(O)=O | |
156.07 | |
156.07 | |
Sodium hydrogen DL-malate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only