missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydrogen selenite, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1217330

 View more versions of this product

Catalog No. AAA1217330


Explore available promotions

Description

Description

Sodium hydrogen selenite is used as a culture media additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen selenite is used as a culture media additive.

Solubility
Soluble in water.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Specifications

Specifications

Sodium hydrogen selenite
250 g
MFCD00003525
sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide
BVTBRVFYZUCAKH-UHFFFAOYSA-L
disodium selenite
23669629
98%
7782-82-3
Na2O3Se
UN2630
Soluble in water.
[Na+].[Na+].[O-][Se]([O-])=O
172.95
150.96
Hygroscopic

Chemical Identifiers

7782-82-3
172.95
BVTBRVFYZUCAKH-UHFFFAOYSA-L
23669629
Na2O3Se
MFCD00003525
sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide
[Na+].[Na+].[O-][Se]([O-])=O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c

H300-H331-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES

EINECSNumber : 231-966-3

RTECSNumber : VS7500000

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only