missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydrogen selenite, 96%, Thermo Scientific Chemicals

$96.14 - $363.56

Chemical Identifiers

CAS 7782-82-3
Molecular Formula Na2O3Se
Molecular Weight (g/mol) 172.95
MDL Number MFCD00003525
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L
Synonym sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide
PubChem CID 23669629
SMILES [Na+].[Na+].[O-][Se]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1217318
View Documents
Thermo Scientific Chemicals
A1217318
50 g
Each for $96.14
 
AAA1217330
View Documents
Thermo Scientific Chemicals
A1217330
250 g
Each for $363.56
 
Description

Description

Sodium hydrogen selenite is used as a culture media additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen selenite is used as a culture media additive.

Solubility
Soluble in water.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

7782-82-3
172.95
BVTBRVFYZUCAKH-UHFFFAOYSA-L
23669629
Na2O3Se
MFCD00003525
sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide
[Na+].[Na+].[O-][Se]([O-])=O

Specifications

7782-82-3
Na2O3Se
UN2630
Soluble in water.
[Na+].[Na+].[O-][Se]([O-])=O
172.95
150.96
Hygroscopic
50 g
MFCD00003525
sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide
BVTBRVFYZUCAKH-UHFFFAOYSA-L
sodium;hydrogen selenite
23669629
98%
Sodium hydrogen selenite
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c

H300-H331-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES

EINECSNumber : 231-966-3

RTECSNumber : VS7500000

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only