missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydrosulfite (Powder/Laboratory), Fisher Chemical™
$250.54 - $250.54
Chemical Identifiers
| CAS | 7775-14-6 |
|---|---|
| Molecular Formula | Na2O4S2 |
| Molecular Weight (g/mol) | 174.096 |
| MDL Number | MFCD00011640 |
| InChI Key | JVBXVOWTABLYPX-UHFFFAOYSA-L |
| Synonym | sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros |
| PubChem CID | 24489 |
| ChEBI | CHEBI:66870 |
| SMILES | [O-]S(=O)S(=O)[O-].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
S310-100
|
Thermo Fisher Scientific
S310100 |
100 g | Glass Bottle |
Each for $250.54
Case of 6 Each for $1,503.24
|
|
||||
Chemical Identifiers
| 7775-14-6 | |
| 174.096 | |
| JVBXVOWTABLYPX-UHFFFAOYSA-L | |
| 24489 | |
| [O-]S(=O)S(=O)[O-].[Na+].[Na+] |
| Na2O4S2 | |
| MFCD00011640 | |
| sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros | |
| CHEBI:66870 |
Specifications
| 300°C | |
| 7775-14-6 | |
| Powder/Solid | |
| Na2O4S2 | |
| sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros | |
| [O-]S(=O)S(=O)[O-].[Na+].[Na+] | |
| 24489 | |
| 174.1 | |
| Pass Test | |
| 1.4g/cm³ |
| White | |
| 8 to 10.5 | |
| 100 g | |
| MFCD00011640 | |
| JVBXVOWTABLYPX-UHFFFAOYSA-L | |
| 174.096 | |
| CHEBI:66870 | |
| Laboratory | |
| Glass Bottle | |
| Sodium Hydrosulfite |
Safety and Handling
DOTInformation : DOT Class 4.2, : Spontaneously Combustible