missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydrosulfite (Powder/Laboratory), Fisher Chemical™

Supplier:  Thermo Fisher Scientific S310100

 View more versions of this product

Catalog No. S310-100



Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hydrosulfite
White
8 to 10.5
100 g
MFCD00011640
JVBXVOWTABLYPX-UHFFFAOYSA-L
174.096
CHEBI:66870
Laboratory
Glass Bottle
300°C
7775-14-6
Powder/Solid
Na2O4S2
sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
[O-]S(=O)S(=O)[O-].[Na+].[Na+]
24489
174.1
Pass Test
1.4g/cm³

Chemical Identifiers

7775-14-6
174.096
JVBXVOWTABLYPX-UHFFFAOYSA-L
24489
[O-]S(=O)S(=O)[O-].[Na+].[Na+]
Na2O4S2
MFCD00011640
sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros
CHEBI:66870
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 4.2, : Spontaneously Combustible

SDS Documents

Documents

Promotions

Promotions