Learn More
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 424330025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium hydroxide | |
1390.0°C | |
318.0°C | |
HNaO | |
01,1083; 05,616; 07,336; 08,460 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
2.5 kg | |
[OH-].[Na+] | |
40 | |
CHEBI:32145 | |
≥97% | |
Plastic Bottle | |
Pellet |
White | |
1310-73-2 | |
97+% | |
NaOH | |
15, 8761 | |
Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol,insoluble in acetone | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
ACS Reagent | |
1mbar at 700°C |
Chemical Identifiers
1310-73-2 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 215-185-5
RUO – Research Use Only