missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
$131.97 - $737.28
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC424335000
|
Thermo Scientific Chemicals
424335000 |
500 g |
Each for $131.97
Case of 6 Each for $0.00
|
|
|||||
AC424330010
|
Thermo Scientific Chemicals
424330010 |
1 kg |
Each for $240.00
|
|
|||||
AC424330025
|
Thermo Scientific Chemicals
424330025 |
2.5 kg |
Each for $295.19
Case of 4 Each for $0.00
|
|
|||||
AC424330100
|
Thermo Scientific Chemicals
424330100 |
10 kg |
Each for $737.28
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1310-73-2 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
White | |
1310-73-2 | |
97+% | |
NaOH | |
15, 8761 | |
500 g | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
≥97% | |
Plastic bottle | |
Pellet |
1390.0°C | |
318.0°C | |
HNaO | |
01,1083; 05,616; 07,336; 08,460 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
ACS Reagent | |
1mbar at 700°C | |
Sodium hydroxide |
Safety and Handling
GHS P Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
Emergency Overview
Causes severe burns by all exposure routes. Hygroscopic.
NFPA
Health: 3
Flammability:
Danger
RUO – Research Use Only