Learn More
Sodium hydroxide, flake, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1839536
Description
Sodium hydroxide flake is widely used in a variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as in water treatment plants. It is used in the Bayer process of aluminum manufacture as well as to remove sulfurous impurities from crude oil. It is also used in the manufacture of biodiesel, rayon and cellophane; as a catalyst for the trans esterification of methanol and triglycerides and as paint stripper on wooden objects. It is mainly used for pH control, acid neutralization, off-gas scrubbing and as a catalyst in various industries like pharmaceutical, agrochemical, dyes, paints and polymer. It is also used in refining vegetable oils, rubber reclamation, metal processing, metal degreasing, adhesive preparations, paint removers, disinfectants, rubber latex stabilizers and in the stabilization of sodium hypochlorite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium hydroxide flake is widely used in a variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as in water treatment plants. It is used in the Bayer process of aluminum manufacture as well as to remove sulfurous impurities from crude oil. It is also used in the manufacture of biodiesel, rayon and cellophane; as a catalyst for the trans esterification of methanol and triglycerides and as paint stripper on wooden objects. It is mainly used for pH control, acid neutralization, off-gas scrubbing and as a catalyst in various industries like pharmaceutical, agrochemical, dyes, paints and polymer. It is also used in refining vegetable oils, rubber reclamation, metal processing, metal degreasing, adhesive preparations, paint removers, disinfectants, rubber latex stabilizers and in the stabilization of sodium hypochlorite.
Notes
Air sensitive and hygroscopic. It is deliquescent and readily absorbs moisture and carbon dioxide in air. Incompatible with strong oxidizing agents, strong acids and organic materials.
Specifications
14 | |
1390°C | |
318°C | |
MFCD00003548 | |
2.13 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
500 g | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
98% | |
Flakes |
Sodium hydroxide | |
1310-73-2 | |
HNaO | |
UN1823 | |
14,8627 | |
Soluble in water,methanol,glycerol and ethanol. Insoluble in acetone and ether. | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
Odorless |
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Safety and Handling
GHS H Statement
H314
Causes severe skin burns and eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM HYDROXIDE, SOLID
EINECSNumber : 215-185-5
RTECSNumber : WB4900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only