missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydroxide, flake, 98%, Thermo Scientific Chemicals

$60.53 - $543.81

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1839536
View Documents
Thermo Scientific Chemicals
A1839536
500 g
Each for $60.53
 
AAA183950E
View Documents
Thermo Scientific Chemicals
A183950E
2500 g
Each for $177.51
 
AAA183950C
View Documents
Thermo Scientific Chemicals
A183950C
10,000 g
Each for $543.81
 
Description

Description

Sodium hydroxide flake is widely used in a variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as in water treatment plants. It is used in the Bayer process of aluminum manufacture as well as to remove sulfurous impurities from crude oil. It is also used in the manufacture of biodiesel, rayon and cellophane; as a catalyst for the trans esterification of methanol and triglycerides and as paint stripper on wooden objects. It is mainly used for pH control, acid neutralization, off-gas scrubbing and as a catalyst in various industries like pharmaceutical, agrochemical, dyes, paints and polymer. It is also used in refining vegetable oils, rubber reclamation, metal processing, metal degreasing, adhesive preparations, paint removers, disinfectants, rubber latex stabilizers and in the stabilization of sodium hypochlorite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydroxide flake is widely used in a variety of large scale applications such as pulp and paper manufacturing, in the production of soap and detergents as well as in water treatment plants. It is used in the Bayer process of aluminum manufacture as well as to remove sulfurous impurities from crude oil. It is also used in the manufacture of biodiesel, rayon and cellophane; as a catalyst for the trans esterification of methanol and triglycerides and as paint stripper on wooden objects. It is mainly used for pH control, acid neutralization, off-gas scrubbing and as a catalyst in various industries like pharmaceutical, agrochemical, dyes, paints and polymer. It is also used in refining vegetable oils, rubber reclamation, metal processing, metal degreasing, adhesive preparations, paint removers, disinfectants, rubber latex stabilizers and in the stabilization of sodium hypochlorite.

Notes
Air sensitive and hygroscopic. It is deliquescent and readily absorbs moisture and carbon dioxide in air. Incompatible with strong oxidizing agents, strong acids and organic materials.
Specifications

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]

Specifications

14
1310-73-2
HNaO
UN1823
14,8627
Soluble in water,methanol,glycerol and ethanol. Insoluble in acetone and ether.
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Odorless
Sodium hydroxide
1390°C
318°C
MFCD00003548
2.13
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
500 g
[OH-].[Na+]
39.997
CHEBI:32145
98%
Flakes
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM HYDROXIDE, SOLID

EINECSNumber : 215-185-5

RTECSNumber : WB4900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only