missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite Solution (4-6%/Laboratory), Fisher Chemical™

$228.42 - $429.83

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
SS290-1 Edge
SDS View Documents Promotion Details
Thermo Fisher Scientific
SS2901
1 L Poly Bottle
Each for $228.42
Case of 6 Each for $1,370.52
Only null left
Edge
 
SS290-4
SDS View Documents Promotion Details
Fisher Chemical
SS2904
4 L Poly Bottle
Each for $429.83
Case of 4 Each for $1,719.32
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7681-52-9
74.439
SUKJFIGYRHOWBL-UHFFFAOYSA-N
23665760
sodium;hypochlorite
ClNaO
MFCD00011120
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
CHEBI:32146
[O-]Cl.[Na+]

Specifications

Yellow
0°C
MFCD00011120
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
[O-]Cl.[Na+]
74.439
CHEBI:32146
4 to 6%
Pass Test
14mmHg
Liquid
7681-52-9,7732-18-5
ClNaO
1.1g/cm³
SUKJFIGYRHOWBL-UHFFFAOYSA-N
sodium;hypochlorite
23665760
74.44
Laboratory
Poly Bottle
1 L
Sodium Hypochlorite Solution, 4-6%
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS
missing translation for 'documents'

missing translation for 'documents'