missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hypochlorite Solution (4-6%/Laboratory), Fisher Chemical™
Supplier: Fisher Chemical SS2904
Specifications
Sodium Hypochlorite Solution | |
4-6% | |
0°C | |
MFCD00011120 | |
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
[O-]Cl.[Na+] | |
74.439 | |
CHEBI:32146 | |
4 to 6% | |
Pass Test | |
14mmHg | |
Liquid |
Yellow | |
7681-52-9,7732-18-5 | |
ClNaO | |
1.1g/cm³ | |
SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
sodium;hypochlorite | |
23665760 | |
74.44 | |
Laboratory | |
Poly Bottle | |
4 L |
Chemical Identifiers
7681-52-9 | |
74.439 | |
SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
23665760 | |
sodium;hypochlorite |
ClNaO | |
MFCD00011120 | |
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution | |
CHEBI:32146 | |
[O-]Cl.[Na+] |
Safety and Handling
DOTInformation : DOT Class 8, : Corrosive