Learn More
Sodium ingot, 99.8% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01034235
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver to Gray | |
Product meets ACS specifications | |
Silver Gray ingot | |
99.8% (Metals basis) | |
MFCD00085307 | |
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot | |
[Na] | |
22.99 | |
CHEBI:26708 | |
Odorless | |
Ingot |
Sodium | |
7440-23-5 | |
0.968g/cm3 | |
Na | |
450 g | |
KEAYESYHFKHZAL-UHFFFAOYSA-N | |
sodium | |
5360545 | |
ACS | |
1.59 hPa (1mm Hg) |
Chemical Identifiers
7440-23-5 | |
22.99 | |
KEAYESYHFKHZAL-UHFFFAOYSA-N | |
5360545 | |
sodium |
Na | |
MFCD00085307 | |
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot | |
CHEBI:26708 | |
[Na] |
Safety and Handling
P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
H260-H314-H335
DOTInformation : DOT Class: 4.3, Packaging Group: I
EINECSNumber : 231-132-9
RTECSNumber : VY0686000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only