missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium ingot, 99.8% (metals basis), Thermo Scientific Chemicals

$239.23 - $721.34

Chemical Identifiers

CAS 7440-23-5
Molecular Formula Na
Molecular Weight (g/mol) 22.99
MDL Number MFCD00085307
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
Synonym natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
PubChem CID 5360545
ChEBI CHEBI:26708
IUPAC Name sodium
SMILES [Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1034235
View Documents
Thermo Scientific Chemicals
01034235
450 g
Each for $239.23
 
AA1034260
View Documents
Thermo Scientific Chemicals
01034260
5 x 450 g
Each for $721.34
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-23-5
22.99
KEAYESYHFKHZAL-UHFFFAOYSA-N
5360545
sodium
Na
MFCD00085307
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
CHEBI:26708
[Na]

Specifications

Silver to Gray
Silver Gray ingot
99.8% (Metals basis)
MFCD00085307
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
[Na]
22.99
CHEBI:26708
Odorless
Ingot
7440-23-5
0.968g/cm3
Na
450 g
KEAYESYHFKHZAL-UHFFFAOYSA-N
sodium
5360545
ACS
1.59 hPa (1mm Hg)
Sodium, Product meets ACS specifications
Videos
Safety and Handling

Safety and Handling

P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H260-H314-H335

DOTInformation : DOT Class: 4.3, Packaging Group: I

EINECSNumber : 231-132-9

RTECSNumber : VY0686000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only